Dimetylftalat 95%
Leverantör: Apollo Scientific
Synonyms: Phthalic acid dimethyl ester
OR922685-100G OR922685-1KG OR922685-500G
APOSOR922685-100GEA 585 SEK
APOSOR922685-100G APOSOR922685-1KG APOSOR922685-500G
Dimetylftalat 95%
Dimetylftalat
Formel: C₁₀H₁₀O₄
MW: 194,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00008425
CAS nummer: 131-11-3