4',5'-Dimethyl-2'-methoxyacetophenone 95%
Leverantör: Apollo Scientific
OR922611-1G OR922611-5G
APOSOR922611-1GEA 1975 SEK
APOSOR922611-1G APOSOR922611-5G
4',5'-Dimethyl-2'-methoxyacetophenone 95%
4',5'-Dimethyl-2'-methoxyacetophenone
Formel: C₁₁H₁₄O₂
Storage Temperature: Omgivande
MDL Number: MFCD03789142
CAS nummer: 91969-74-3