4-Amylcyclohexanol (cis- and trans- mixture) 95%
Leverantör: Apollo Scientific
OR922544-25G OR922544-5G
APOSOR922544-25GEA 645 SEK
APOSOR922544-25G APOSOR922544-5G
4-Amylcyclohexanol (cis- and trans- mixture) 95%
4-Amylcyclohexanol (cis- and trans- mixture)
Formel: C₁₁H₂₂O
Storage Temperature: Omgivande
MDL Number: MFCD01017412
CAS nummer: 54410-90-1