2,3-Dimethylcyclohexanol (mixture of isomers) 98%
Leverantör: Apollo Scientific
Synonyms: 2,3-Dimethylcyclohexanol
OR922265-10G OR922265-1G OR922265-5G
APOSOR922265-10GEA 4875 SEK
APOSOR922265-10G APOSOR922265-1G APOSOR922265-5G
2,3-Dimethylcyclohexanol (mixture of isomers) 98%
2,3-Dimethylcyclohexanol (mixture of isomers)
Formel: C₈H₁₆O
Kokpunkt: 181 °C
Densitet: 0,93 g/cm³ (25 °C)
Flampunkt: 66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001444
CAS nummer: 1502-24-5