5-Methyl-2-furonitrile 95%

Leverantör: Apollo Scientific

Synonyms: 5-Methylfuran-2-carbonitrile

OR922196-1G OR922196-250MG
APOSOR922196-1GEA 3875 SEK
APOSOR922196-1G APOSOR922196-250MG
5-Methyl-2-furonitrile 95%
5-Methyl-2-furonitrile
Formel: C₆H₅NO
MW: 107,11 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02093782
CAS nummer: 13714-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR