1-Nitro-2-naphthol 95%
Leverantör: Apollo Scientific
Synonyms:
2-Hydroxy-1-nitronaphthalene, 1-Nitronaphthalen-2-ol
OR922194-1G
OR922194-250MG
APOSOR922194-1GEA
5400
SEK
APOSOR922194-1G
APOSOR922194-250MG
1-Nitro-2-naphthol 95%
1-Nitro-2-naphthol
Formel:
C₁₀H₇NO₃ MW: 189,17 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00021461 CAS nummer: 550-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...