2-Isopropoxyethyl methanesulphonate 95%

Leverantör: Apollo Scientific

Synonyms: MEthanesulphonic acid 2-isopropoxyethyl ester, Methanesulphonic acid 2-Isopropoxyethyl Ester

OR922037-1G OR922037-25G OR922037-5G
APOSOR922037-1GEA 925 SEK
APOSOR922037-1G APOSOR922037-25G APOSOR922037-5G
2-Isopropoxyethyl methanesulphonate 95%
2-Isopropoxyethyl methanesulphonate
Formel: C₆H₁₄O₄S
MW: 182.23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02093478
CAS nummer: 235097-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR