Natriumformiat 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
Formic acid sodium salt
OR921745-5KG
OR921745-1KG
APOSOR921745-5KGEA
1725
SEK
APOSOR921745-5KG
APOSOR921745-1KG
Natriumformiat 98%
Natriumformiat
Formel:
HCO₂Na MW: 68,01 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00013101 CAS nummer: 141-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...