3-Methylpentyl acetate 95%
Leverantör: Apollo Scientific
Synonyms:
Acetic Acid 3-Methylpentyl Ester
OR921375-25G
OR921375-5G
APOSOR921375-25GEA
5050
SEK
APOSOR921375-25G
APOSOR921375-5G
3-Methylpentyl acetate 95%
3-Methylpentyl acetate
Formel:
C₈H₁₆O₂ MW: 144.21 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00671487 CAS nummer: 35897-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...