Cycloheptanone oxime 95%

Leverantör: Apollo Scientific

Synonyms: Suberoxime

OR921124-1G OR921124-25G OR921124-5G
APOSOR921124-1GEA 1550 SEK
APOSOR921124-1G APOSOR921124-25G APOSOR921124-5G
Cycloheptanone oxime 95%
Cycloheptanone oxime
Formel: C₇H₁₃NO
MW: 127,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00046508
CAS nummer: 2158-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR