1,2-Di(4-pyridyl)ethane 95%
Leverantör: Apollo Scientific
Synonyms: 4,4'-Ethylenedipyridine, 1,2-Di(pyridin-4-yl)ethane, 1,2-Bis(4-pyridyl)ethane
OR920918-1G OR920918-5G
APOSOR920918-1GEA 925 SEK
APOSOR920918-1G APOSOR920918-5G
1,2-Di(4-pyridyl)ethane 95%
1,2-Di(4-pyridyl)ethane
Formel: C₁₂H₁₂N₂
MW: 184,24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00006451
CAS nummer: 4916-57-8