1,2-Di(4-pyridyl)ethylene 98+%
Leverantör: Apollo Scientific
Synonyms: (E)-1,2-Di(pyridin-4-yl)ethene, 1,2-Bis(4-pyridyl)ethylene, 4,4'-Vinylenedipyridine
OR920301-1G OR920301-5G
APOSOR920301-1GEA 720 SEK
APOSOR920301-1G APOSOR920301-5G
1,2-Di(4-pyridyl)ethylene 98+%
1,2-Di(4-pyridyl)ethylene
Formel: C₁₂H₁₀N₂
MW: 182,23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00006448
CAS nummer: 13362-78-2