(Bromomethyl)triphenylphosphonium bromide 97%
Leverantör: Apollo Scientific
Varning
OR920209-100G OR920209-25G OR920209-5G
APOSOR920209-100GEA 5600 SEK
APOSOR920209-100G APOSOR920209-25G APOSOR920209-5G
(Bromomethyl)triphenylphosphonium bromide 97%
(Bromomethyl)triphenylphosphonium bromide
Formel: C₁₉H₁₇Br₂P
MW: 436,1200 g/mol
Smältpunkt: 234…236 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011864
CAS nummer: 1034-49-7