Benzylmalonic acid 98%
Leverantör: Apollo Scientific
OR920047-25G OR920047-5G
APOSOR920047-25GEA 1300 SEK
APOSOR920047-25G APOSOR920047-5G
Benzylmalonic acid 98%
Benzylmalonic acid
Formel: C₁₀H₁₀O₄
Storage Temperature: Omgivande
MDL Number: MFCD00004261
CAS nummer: 616-75-1