2-(2,6-Dichlorophenyl)ethanol 95%
Leverantör: Apollo Scientific
Synonyms: 2,6-Dichlorophenethyl Alcohol
OR919967-1G OR919967-25G OR919967-5G
APOSOR919967-1GEA 1475 SEK
APOSOR919967-1G APOSOR919967-25G APOSOR919967-5G
2-(2,6-Dichlorophenyl)ethanol 95%
2-(2,6-Dichlorophenyl)ethanol
Formel: C₈H₈Cl₂O
MW: 191,05 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00800674
CAS nummer: 30595-79-0