4-Hydroxyphthalonitrile 95%

Leverantör: Apollo Scientific

Synonyms: 3,4-Dicyanophenol

OR919916-1G OR919916-5G
APOSOR919916-1GEA 1025 SEK
APOSOR919916-1G APOSOR919916-5G
4-Hydroxyphthalonitrile 95%
4-Hydroxyphthalonitrile
Formel: C₈H₄N₂O
MW: 144,13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00191653
CAS nummer: 30757-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR