Vinyl cinnamate 95%

Leverantör: Apollo Scientific

Synonyms: Cinnamic Acid Vinyl Ester

OR919498-100G OR919498-25G OR919498-500G
APOSOR919498-100GEA 4725 SEK
APOSOR919498-100G APOSOR919498-25G APOSOR919498-500G
Vinyl cinnamate 95%
Vinyl cinnamate
Formel: C₁₁H₁₀O₂
MW: 174,20 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00059324
CAS nummer: 3098-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR