4-Cyclohexylcyclohexanol 95%
Leverantör: Apollo Scientific
OR919464-1G OR919464-25G OR919464-5G
APOSOR919464-1GEA 1375 SEK
APOSOR919464-1G APOSOR919464-25G APOSOR919464-5G
4-Cyclohexylcyclohexanol 95%
4-Cyclohexylcyclohexanol
Formel: C₁₂H₂₂O
Storage Temperature: Omgivande
MDL Number: MFCD00060071
CAS nummer: 2433-14-9