4-Aminobenzophenone 98%
Leverantör: Apollo Scientific
Synonyms: (4-Aminophenyl)(phenyl)methanone, p-Aminobenzophenone
OR919301-25G OR919301-5G OR919301-100G
APOSOR919301-25GEA 1125 SEK
APOSOR919301-25G APOSOR919301-5G APOSOR919301-100G
4-Aminobenzophenone 98%
4-Aminobenzophenone
Formel: C₁₃H₁₁NO
MW: 197,24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007895
CAS nummer: 1137-41-3