2-(1,3-Dioxolan-2-yl)furan 98%
Leverantör: Apollo Scientific
Synonyms: Furfural Ethylene Acetal, 2-(2-Furyl)-1,3-dioxolane
OR919261-1G OR919261-25G OR919261-5G
APOSOR919261-1GEA 1300 SEK
APOSOR919261-1G APOSOR919261-25G APOSOR919261-5G
2-(1,3-Dioxolan-2-yl)furan 98%
2-(1,3-Dioxolan-2-yl)furan
Formel: C₇H₈O₃
MW: 140,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00671520
CAS nummer: 1708-41-4