3-(1,3,5-Trimethyl-1H-pyrazol-4-yl)propanoic acid 95%
Leverantör: Apollo Scientific
OR919056-1G OR919056-5G
APOSOR919056-1GEA 2325 SEK
APOSOR919056-1G APOSOR919056-5G
3-(1,3,5-Trimethyl-1H-pyrazol-4-yl)propanoic acid 95%
3-(1,3,5-Trimethyl-1H-pyrazol-4-yl)propanoic acid
Formel: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04970957
CAS nummer: 956352-96-8