N-Isopropylacrylamide 95%
Leverantör: Apollo Scientific
OR918836-25G
OR918836-100G
OR918836-500G
APOSOR918836-25GEA
945
SEK
APOSOR918836-25G
APOSOR918836-100G
APOSOR918836-500G
N-Isopropylacrylamide 95%
N-Isopropylacrylamide
Formel:
C₆H₁₁NO MW: 113,16 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00041913 CAS nummer: 2210-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...