Methyl-3-benzoylpropionate 97%

Leverantör: Apollo Scientific

Synonyms: Methyl 4-Oxo-4-phenylbutyrate, 3-Benzoylpropionic Acid Methyl Ester, Methyl 4-oxo-4-phenylbutanoate, 4-Oxo-4-phenylbutyric Acid Methyl Ester

OR918821-25G OR918821-5G
APOSOR918821-25GEA 1750 SEK
APOSOR918821-25G APOSOR918821-5G
Methyl-3-benzoylpropionate 97%
Methyl-3-benzoylpropionate
Formel: C₁₁H₁₂O₃
MW: 192,21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00014906
CAS nummer: 25333-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR