2-(2-Methoxyphenyl)ethanol 97%
Leverantör: Apollo Scientific
OR918735-10G OR918735-1G OR918735-5G
APOSOR918735-10GEA 3625 SEK
APOSOR918735-10G APOSOR918735-1G APOSOR918735-5G
2-(2-Methoxyphenyl)ethanol 97%
2-(2-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00004512
CAS nummer: 7417-18-7