N-Methyl-3-chloropropylamine hydrochloride 95%

Leverantör: Apollo Scientific

Synonyms: 3-(Methylamino)propyl Chloride Hydrochloride, 3-Chloro-N-methylpropan-1-amine hydrochloride

OR918572-1G OR918572-5G
APOSOR918572-1GEA 635 SEK
APOSOR918572-1G APOSOR918572-5G
N-Methyl-3-chloropropylamine hydrochloride 95%
N-Methyl-3-chloropropylamine hydrochloride
Formel: C₄H₁₀ClN
MW: 144,04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00144935
CAS nummer: 97145-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR