N-Methyl-1-(tetrahydro-2H-pyran-2-yl)methanamine 95%
Leverantör: Apollo Scientific
OR918463-1G OR918463-250MG
APOSOR918463-1GEA 3175 SEK
APOSOR918463-1G APOSOR918463-250MG
N-Methyl-1-(tetrahydro-2H-pyran-2-yl)methanamine 95%
N-Methyl-1-(tetrahydro-2H-pyran-2-yl)methanamine
Formel: C₇H₁₅NO
MW: 129.2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06738978
CAS nummer: 7179-96-6