Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)isocyanurate 96%
Leverantör: Apollo Scientific
Synonyms: Isocyanuric Acid Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl) Ester
OR918454-100G OR918454-25G OR918454-5G
APOSOR918454-100GEA 0 SEK
APOSOR918454-100G APOSOR918454-25G APOSOR918454-5G
Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)isocyanurate 96%
Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)isocyanurate
Formel: C₄₂H₅₇N₃O₆
MW: 699.93 g/mol
Smältpunkt: 163…165 °C
Storage Temperature: Omgivande
MDL Number: MFCD01026105
CAS nummer: 40601-76-1