2-(1H-Pyrazol-1-ylmethyl)benzaldehyde 95%
Leverantör: Apollo Scientific
OR918319-1G OR918319-250MG
APOSOR918319-1GEA 4700 SEK
APOSOR918319-1G APOSOR918319-250MG
2-(1H-Pyrazol-1-ylmethyl)benzaldehyde 95%
2-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Formel: C₁₁H₁₀N₂O
Storage Temperature: Omgivande
MDL Number: MFCD09864294
CAS nummer: 1177332-23-8