1,3-Di(4-pyridyl)propane 95%
Leverantör: Apollo Scientific
Synonyms: 4,4'-Trimethylenedipyridine
OR918123-100G OR918123-25G
APOSOR918123-100GEA 4050 SEK
APOSOR918123-100G APOSOR918123-25G
1,3-Di(4-pyridyl)propane 95%
1,3-Di(4-pyridyl)propane
Formel: C₁₃H₁₄N₂
MW: 198,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00038046
CAS nummer: 17252-51-6