Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 4-bromo-1-methyl-1H-pyrazole-3-carboxylate
OR918037-1G OR918037-250MG OR918037-5G
APOSOR918037-1GEA 2250 SEK
APOSOR918037-1G APOSOR918037-250MG APOSOR918037-5G
Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate 98%
Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate
Formel: C₆H₇BrN₂O₂
MW: 219,04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02253785
CAS nummer: 211738-66-8