1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 97%
Leverantör: Apollo Scientific
Synonyms: Cyclohexane-1,3-dicarboxylic acid, 1,3-Cyclohexanedicarboxylic acid
OR917608-25G OR917608-100G
APOSOR917608-25GEA 1075 SEK
APOSOR917608-25G APOSOR917608-100G
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 97%
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)
Formel: C₈H₁₂O₄
MW: 172,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00134411
CAS nummer: 3971-31-1