2,3-Epoxy-1-(1-ethoxyethoxy)propane 95%
Leverantör: Apollo Scientific
Synonyms: 2-((1-Ethoxyethoxy)methyl)oxirane
OR917210-5G OR917210-25G
APOSOR917210-5GEA 2300 SEK
APOSOR917210-5G APOSOR917210-25G
2,3-Epoxy-1-(1-ethoxyethoxy)propane 95%
2,3-Epoxy-1-(1-ethoxyethoxy)propane
Formel: C₇H₁₄O₃
MW: 146,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22575236
CAS nummer: 4416-85-7