4-Bromodibenzothiophene 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Bromodibenzo[b,d]thiophene
OR916957-25G
OR916957-50G
APOSOR916957-25GEA
3025
SEK
APOSOR916957-25G
APOSOR916957-50G
4-Bromodibenzothiophene 98%
4-Bromodibenzothiophene
Formel:
C₁₂H₇BrS MW: 263,15 g/mol Smältpunkt: 87 °C Storage Temperature: Omgivande |
MDL Number:
MFCD02683746 CAS nummer: 97511-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...