3-(4-Methoxyphenyl)propionic acid 98%
Leverantör: Apollo Scientific
Synonyms: 3-(4-Methoxyphenyl)propanoic acid
OR914551-100G OR914551-500G
APOSOR914551-100GEA 1075 SEK
APOSOR914551-100G APOSOR914551-500G
3-(4-Methoxyphenyl)propionic acid 98%
3-(4-Methoxyphenyl)propionic acid
Formel: C₁₀H₁₂O₃
MW: 180.2 g/mol
Kokpunkt: 319
Storage Temperature: Omgivande
MDL Number: MFCD00002777
CAS nummer: 1929-29-9