Boc-3-bromo-L-phenylalanine 98%

Leverantör: Apollo Scientific

Synonyms: Boc-Phe(3-Br)-OH

OR914325-1G OR914325-5G
APOSOR914325-1GEA 1350 SEK
APOSOR914325-1G APOSOR914325-5G
Boc-3-bromo-L-phenylalanine 98%
Boc-3-bromo-L-phenylalanine
Formel: C₁₄H₁₈BrNO₄
MW: 344,21 g/mol
Smältpunkt: 130…140 °C
Storage Temperature: Omgivande
MDL Number: MFCD01317710
CAS nummer: 82278-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR