2-Bromo-4,6-dimethylpyridine 98%
Leverantör: Apollo Scientific
OR914244-10G OR914244-1G OR914244-5G
APOSOR914244-10GEA 8475 SEK
APOSOR914244-10G APOSOR914244-1G APOSOR914244-5G
2-Bromo-4,6-dimethylpyridine 98%
2-Bromo-4,6-dimethylpyridine
Formel: C₇H₈BrN
MW: 186,05 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00082591
CAS nummer: 4926-26-5