(S)-3-Amino-3-(3-nitrophenyl)propionic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (S)-3-Amino-3-(3-nitrophenyl)propanoic acid
OR913926-1G OR913926-250MG OR913926-5G
APOSOR913926-1GEA 2100 SEK
APOSOR913926-1G APOSOR913926-250MG APOSOR913926-5G
(S)-3-Amino-3-(3-nitrophenyl)propionic acid 97%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid
Formel: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04113693
CAS nummer: 734529-57-8