(R)-(+)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-N-(Methoxymethyl)-1-phenyl-N-((trimethylsilyl)methyl)ethanamine
OR913189-1G OR913189-5G
APOSOR913189-1GEA 3000 SEK
APOSOR913189-1G APOSOR913189-5G
(R)-(+)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine 97%
(R)-(+)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
Formel: C₁₄H₂₅NOSi
MW: 251.4399 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06798959
CAS nummer: 133407-38-2