(S)-1-(2-Chlorophenyl)ethanol 95%

Leverantör: Apollo Scientific

OR913172-1G OR913172-250MG
APOSOR913172-1GEA 1825 SEK
APOSOR913172-1G APOSOR913172-250MG
(S)-1-(2-Chlorophenyl)ethanol 95%
(S)-1-(2-Chlorophenyl)ethanol
Formel: C₈H₉ClO
MW: 156,609 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06659506
CAS nummer: 131864-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR