2-Methyl-3H-imidazo[4,5-b]pyridine hydrochloride 98%
Leverantör: Apollo Scientific
Synonyms: 2-Methyl-3H-imidazo[4,5-b]pyridine
OR912899-1G OR912899-5G
APOSOR912899-1GEA 1600 SEK
APOSOR912899-1G APOSOR912899-5G
2-Methyl-3H-imidazo[4,5-b]pyridine hydrochloride 98%
2-Methyl-3H-imidazo[4,5-b]pyridine hydrochloride
Formel: C₇H₈ClN₃
MW: 169,61 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16039350
CAS nummer: 68175-07-5