3,5,5-Trimethylcyclohexane-1,2-dione 95%

Leverantör: Apollo Scientific

OR912808-10G OR912808-1G OR912808-5G
APOSOR912808-10GEA 925 SEK
APOSOR912808-10G APOSOR912808-1G APOSOR912808-5G
3,5,5-Trimethylcyclohexane-1,2-dione 95%
3,5,5-Trimethylcyclohexane-1,2-dione
Formel: C₉H₁₄O₂
MW: 154,209 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02181023
CAS nummer: 57696-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR