4-Chloro-N-methoxy-N-methylbenzamide 95%

Leverantör: Apollo Scientific

OR912801-1G OR912801-5G
APOSOR912801-1GEA 600 SEK
APOSOR912801-1G APOSOR912801-5G
4-Chloro-N-methoxy-N-methylbenzamide 95%
4-Chloro-N-methoxy-N-methylbenzamide
Formel: C₉H₁₀ClNO₂
MW: 199,63 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02684308
CAS nummer: 122334-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR