2-Chloro-4-nitropyridine-N-oxide 98%
Leverantör: Apollo Scientific
Synonyms: 2-Chloro-4-nitropyridine N-oxide
OR912604-100G OR912604-25G
APOSOR912604-100GEA 6175 SEK
APOSOR912604-100G APOSOR912604-25G
2-Chloro-4-nitropyridine-N-oxide 98%
2-Chloro-4-nitropyridine-N-oxide
Formel: C₅H₃ClN₂O₃
MW: 174,54 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00955617
CAS nummer: 14432-16-7