2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone 95%
Leverantör: Apollo Scientific
Synonyms:
2-Ethyl-4-hydroxy-5-methylfuran-3(2H)-one, Ethyl furaneol
OR912424-10G
OR912424-1G
OR912424-5G
APOSOR912424-10GEA
5825
SEK
APOSOR912424-10G
APOSOR912424-1G
APOSOR912424-5G
2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone 95%
2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone
Formel:
C₇H₁₀O₃ MW: 142,15 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00191360 CAS nummer: 27538-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...