1-Ethyl-3-hydroxypiperidine 95%

Leverantör: Apollo Scientific

OR912385-25G OR912385-5G
APOSOR912385-25GEA 705 SEK
APOSOR912385-25G APOSOR912385-5G
1-Ethyl-3-hydroxypiperidine 95%
1-Ethyl-3-hydroxypiperidine
Formel: C₇H₁₅NO
Storage Temperature: Omgivande
MDL Number: MFCD00006508
CAS nummer: 13444-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR