1-Chloro-4-ethoxybenzene 95%
Leverantör: Apollo Scientific
Synonyms:
4-Chlorophenetole, 1-Chloro-4-ethoxybenzene
OR912270-25G
OR912270-5G
APOSOR912270-25GEA
4475
SEK
APOSOR912270-25G
APOSOR912270-5G
1-Chloro-4-ethoxybenzene 95%
1-Chloro-4-ethoxybenzene
Formel:
C₈H₉ClO MW: 156,61 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00058931 CAS nummer: 622-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...