1-Chloro-2-ethoxybenzene 95%

Leverantör: Apollo Scientific

Synonyms: 1-Chloro-2-ethoxybenzene, 2-Chlorophenetole

OR912269-10G OR912269-1G OR912269-5G
APOSOR912269-10GEA 4725 SEK
APOSOR912269-10G APOSOR912269-1G APOSOR912269-5G
1-Chloro-2-ethoxybenzene 95%
1-Chloro-2-ethoxybenzene
Formel: C₈H₉ClO
MW: 156,61 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00058930
CAS nummer: 614-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR