tert-Butyl-3-(methylsulphonyloxymethyl)piperidine-1-carboxylate 97%
Leverantör: Apollo Scientific
Synonyms: 1-Boc-3-Methanesulphonyloxymethyl-piperidine, 1-Boc-3-[(methylsulphonyloxy)methyl]piperidine
OR912177-1G OR912177-25G OR912177-5G
APOSOR912177-1GEA 1525 SEK
APOSOR912177-1G APOSOR912177-25G APOSOR912177-5G
tert-Butyl-3-(methylsulphonyloxymethyl)piperidine-1-carboxylate 97%
tert-Butyl-3-(methylsulphonyloxymethyl)piperidine-1-carboxylate
Formel: C₁₂H₂₃NO₅S
MW: 293,38 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06795963
CAS nummer: 162166-99-6