3-Methylisoquinoline 98%

Leverantör: Apollo Scientific

OR912094-10G OR912094-1G OR912094-5G
APOSOR912094-10GEA 5900 SEK
APOSOR912094-10G APOSOR912094-1G APOSOR912094-5G
3-Methylisoquinoline 98%
3-Methylisoquinoline
Formel: C₁₀H₉N
MW: 143,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00024139
CAS nummer: 1125-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR