2,6-Dimethoxy-p-cresol 95%
Leverantör: Apollo Scientific
Synonyms: 3,5-Dimethoxy-4-hydroxytoluene, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzene, 2,6-Dimethoxy-4-methylphenol
OR912037-100G OR912037-25G OR912037-5G
APOSOR912037-100GEA 9500 SEK
APOSOR912037-100G APOSOR912037-25G APOSOR912037-5G
2,6-Dimethoxy-p-cresol 95%
2,6-Dimethoxy-p-cresol
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00017289
CAS nummer: 6638-05-7